(1S)-1-[4-(Trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride
Catalog # APOSPC200088-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:237.649 g/mol
Storage Temperature:Ambient
CAS Number:1391528-41-8
Formula:C₁₀H₁₀F₃N·ClH
MDL Number:MFCD12068559