Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,5,6-Tetrafluoroisonicotinonitrile
2,3,5,6-Tetrafluoroisonicotinonitrile
2,3,5,6-Tetrafluoroisonicotinonitrile
2,3,5,6-Tetrafluoroisonicotinonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
2,3,5,6-Tetrafluoroisonicotinonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
16297-07-7
:
Formula:
C₆F₄N₂
MDL Number:
MFCD00075302
:
176.071 g/mol
Melting Pt:
66…68 °C
:
GHS07
Storage Temperature:
Ambient