2,6-Difluorophenylhydrazine
Catalog # APOSPC1502-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:1 g
- Seq:3047
Specifications
About this item
:
:144.122 g/mol
Storage Temperature:Raumtemperatur
CAS Number:119452-66-3
Formula:C₆H₆F₂N₂
MDL Number:MFCD06658266