Order Entry
Germany
ContactUsLinkComponent
Diethyl 2-(4-fluorophenyl)malonate
Irritant
Diethyl 2-(4-fluorophenyl)malonate
Catalog # APOSPC10099-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Diethyl 2-(4-fluorophenyl)malonate
Catalog # APOSPC10099-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.254 g/mol
Boiling Pt:161…162 °C (15 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:2965-90-4
Formula:C₁₃H₁₅FO₄
MDL Number:MFCD04039208