
tert-Butyl 4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Catalog # APOSOR9873-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:320.383 g/mol
Melting Pt:165…167 °C
:GHS07
Storage Temperature:Ambient
CAS Number:158985-36-5
Formula:C₁₇H₂₄N₂O₄
MDL Number:MFCD06200865