Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-methyl-1-oxidopyridin-4-yl ether
Methyl 3-methyl-1-oxidopyridin-4-yl ether 96%
Methyl 3-methyl-1-oxidopyridin-4-yl ether 96%
Methyl 3-methyl-1-oxidopyridin-4-yl ether 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 3-methyl-1-oxidopyridin-4-yl ether 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
26883-29-4
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD00234344
:
139.152 g/mol
Storage Temperature:
Ambient