Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Fmoc-N-benzyl-glycine
N-Fmoc-N-benzyl-glycine 97%
N-Fmoc-N-benzyl-glycine 97%
N-Fmoc-N-benzyl-glycine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
N-Fmoc-N-benzyl-glycine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
141743-13-7
:
Formula:
C₂₄H₂₁NO₄
MDL Number:
MFCD04112686
:
387.428 g/mol
Storage Temperature:
Ambient