2-Oxo-2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester 98%
Catalog # APOSOR961194-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:183.204 g/mol
Storage Temperature:Ambient
CAS Number:141293-14-3
Formula:C₉H₁₃NO₃
MDL Number:MFCD01318910