Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(2,4-dihydroxyphenyl)propionate
Methyl 3-(2,4-dihydroxyphenyl)propionate 95%
Methyl 3-(2,4-dihydroxyphenyl)propionate 95%
:
Apollo Scientific
Methyl 3-(2,4-dihydroxyphenyl)propionate 95%
:
Apollo Scientific
:
17422-90-1
:
:
C₁₀H₁₂O₄
:
MFCD19441070
:
196.2 g/mol
:
Ambient