Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 4-hydroxy-3-methyl-1H-pyrazole-1-carboxylate 95%
tert-Butyl 4-hydroxy-3-methyl-1H-pyrazole-1-carboxylate 95%
Catalog # APOSOR957732-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-hydroxy-3-methyl-1H-pyrazole-1-carboxylate 95%
Catalog # APOSOR957732-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.219 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₄N₂O₃
MDL Number:MFCD23110518