Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate 98%
tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate 98%
Catalog # APOSOR957715-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate 98%
Catalog # APOSOR957715-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.192 g/mol
Storage Temperature:Ambient
CAS Number:1394947-77-3
Formula:C₈H₁₂N₂O₃
MDL Number:MFCD23110529