Order Entry
Germany
ContactUsLinkComponent
 
2-Chloro-1-ethoxy-4-methylbenzene 95%
Catalog # APOSOR956963-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-1-ethoxy-4-methylbenzene 95%
Catalog # APOSOR956963-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.636 g/mol
Storage Temperature:Ambient
CAS Number:68758-67-8
Formula:C₉H₁₁ClO
MDL Number:MFCD23725417