Order Entry
Germany
ContactUsLinkComponent
2-Hydroxy-6-(2-methylphenyl)pyridine 96%
2-Hydroxy-6-(2-methylphenyl)pyridine 96%
Catalog # APOSOR954652-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Hydroxy-6-(2-methylphenyl)pyridine 96%
Catalog # APOSOR954652-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.222 g/mol
Storage Temperature:Ambient
CAS Number:300395-32-8
Formula:C₁₂H₁₁NO
MDL Number:MFCD11876169