Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate 95%
Catalog # APOSOR953671-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:217.219 g/mol
Storage Temperature:Freezer
CAS Number:216959-34-1
Formula:C₉H₁₅NO₅
MDL Number:MFCD01631201