Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methyl-2-oxopentanoate
Methyl 4-methyl-2-oxopentanoate 95%
Methyl 4-methyl-2-oxopentanoate 95%
Methyl 4-methyl-2-oxopentanoate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 4-methyl-2-oxopentanoate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
3682-43-7
:
Formula:
C₇H₁₂O₃
MDL Number:
MFCD11035886
:
144.168 g/mol
Storage Temperature:
Ambient