Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Aminomethyl)pyridin-3-ol
2-(Aminomethyl)pyridin-3-ol 97%
2-(Aminomethyl)pyridin-3-ol 97%
APOSOR950277-250MG
:
Apollo Scientific
:
2-(Aminomethyl)pyridin-3-ol 97%
APOSOR950277-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
124.141 g/mol
:
GHS07
:
Refrigerator
:
194665-89-9
:
C₆H₈N₂O
:
MFCD01217253