Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Methoxyphenyl)-1H-pyrazole
1-(4-Methoxyphenyl)-1H-pyrazole 98%
1-(4-Methoxyphenyl)-1H-pyrazole 98%
:
Apollo Scientific
1-(4-Methoxyphenyl)-1H-pyrazole 98%
:
Apollo Scientific
:
35715-67-4
:
:
C₁₀H₁₀N₂O
:
MFCD09033531
:
174.199 g/mol
:
GHS07
:
Ambient