Order Entry
Germany
ContactUsLinkComponent
trans-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
Catalog # APOSOR949332-250MG
Supplier:  Apollo Scientific
CAS Number:  
trans-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
Catalog # APOSOR949332-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :144.168 g/mol
Storage Temperature:Ambient
CAS Number:1408075-48-8
Formula:C₇H₁₂O₃
MDL Number:MFCD23105988