tert-Butyl (2R)-2-(bromomethyl)azetidine-1-carboxylate
Catalog # APOSOR949253-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.133 g/mol
Storage Temperature:Ambient
CAS Number:1363378-13-5
Formula:C₉H₁₆BrNO₂
MDL Number:MFCD22566237