Order Entry
Germany
ContactUsLinkComponent
2-(2-Nitrophenyl)-1,3-dioxolane 95%
2-(2-Nitrophenyl)-1,3-dioxolane 95%
Catalog # APOSOR947260-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Nitrophenyl)-1,3-dioxolane 95%
Catalog # APOSOR947260-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.172 g/mol
Storage Temperature:Ambient
CAS Number:48140-35-8
Formula:C₉H₉NO₄
MDL Number:MFCD06209042