
Methyl 5-aminopentanoate hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29840-56-0
:
Formula:C₆H₁₃NO₂·ClH
MDL Number:MFCD13368250
:167.634 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together