Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,1,5-Trimethyl-1H-pyrazol-3-amine
N,1,5-Trimethyl-1H-pyrazol-3-amine 95%
N,1,5-Trimethyl-1H-pyrazol-3-amine 95%
N,1,5-Trimethyl-1H-pyrazol-3-amine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
N,1,5-Trimethyl-1H-pyrazol-3-amine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
646506-40-3
:
Formula:
C₆H₁₁N₃
MDL Number:
MFCD09991545
:
125.172 g/mol
Storage Temperature:
Ambient