Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methylindole-3-acetonitrile
1-Methylindole-3-acetonitrile 95%
1-Methylindole-3-acetonitrile 95%
1-Methylindole-3-acetonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Methylindole-3-acetonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
51584-17-9
:
Formula:
C₁₁H₁₀N₂
MDL Number:
MFCD09038157
:
170.211 g/mol
Storage Temperature:
Ambient