Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Chloromethyl)furan-2-carbaldehyde
5-(Chloromethyl)furan-2-carbaldehyde 95%
5-(Chloromethyl)furan-2-carbaldehyde 95%
APOSOR939970-1G
:
Apollo Scientific
:
5-(Chloromethyl)furan-2-carbaldehyde 95%
APOSOR939970-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
144.556 g/mol
:
Ambient
:
1623-88-7
:
C₆H₅ClO₂
:
MFCD02181084