Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-phenethylamine hydrochloride
3-Nitro-phenethylamine hydrochloride 97%
3-Nitro-phenethylamine hydrochloride 97%
3-Nitro-phenethylamine hydrochloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Nitro-phenethylamine hydrochloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
19008-62-9
:
Formula:
C₈H₁₀N₂O₂·ClH
MDL Number:
MFCD08059237
:
202.638 g/mol
Storage Temperature:
Ambient