Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
M-Aminobenzyl cyanide
M-Aminobenzyl cyanide 95%
M-Aminobenzyl cyanide 95%
APOSOR930194-1G
:
Apollo Scientific
:
M-Aminobenzyl cyanide 95%
APOSOR930194-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
132.163 g/mol
:
Ambient
:
4623-24-9
:
C₈H₈N₂
:
MFCD03265299