Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-tert-Butoxypropan-1-amine hydrochloride
3-tert-Butoxypropan-1-amine hydrochloride 95%
3-tert-Butoxypropan-1-amine hydrochloride 95%
3-tert-Butoxypropan-1-amine hydrochloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-tert-Butoxypropan-1-amine hydrochloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
864658-14-0
:
Formula:
C₇H₁₇NO·ClH
MDL Number:
MFCD04973192
:
167.677 g/mol
Storage Temperature:
Ambient