Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-[(dimethylamino)methyl]benzoate
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
27171-87-5
:
Formula:
C₁₁H₁₅NO₂
MDL Number:
MFCD13249350
:
193.242 g/mol
Storage Temperature:
Ambient