Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(2-Piperidyl)-1-propanol hydrochloride
3-(2-Piperidyl)-1-propanol hydrochloride 95%
3-(2-Piperidyl)-1-propanol hydrochloride 95%
3-(2-Piperidyl)-1-propanol hydrochloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(2-Piperidyl)-1-propanol hydrochloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
90226-88-3
:
Formula:
C₈H₁₇NO·ClH
MDL Number:
MFCD22573686
:
179.688 g/mol
Storage Temperature:
Ambient