Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Threonine methyl ester hydrochloride
DL-Threonine methyl ester hydrochloride 97%
DL-Threonine methyl ester hydrochloride 97%
DL-Threonine methyl ester hydrochloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
DL-Threonine methyl ester hydrochloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
62076-66-8
:
Formula:
C₅H₁₁NO₃·ClH
MDL Number:
MFCD00070400
:
169.607 g/mol
Storage Temperature:
Refrigerator