Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 1-methyl-1H-pyrazole-3-carboxylate
Methyl 1-methyl-1H-pyrazole-3-carboxylate 98%
Methyl 1-methyl-1H-pyrazole-3-carboxylate 98%
:
Apollo Scientific
Methyl 1-methyl-1H-pyrazole-3-carboxylate 98%
:
Apollo Scientific
:
17827-61-1
:
:
C₆H₈N₂O₂
:
MFCD04967358
:
140.14 g/mol
:
Ambient