Order Entry
Germany
ContactUsLinkComponent
5-Chloropentanal 95%
5-Chloropentanal 95%
Catalog # APOSOR928054-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloropentanal 95%
Catalog # APOSOR928054-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :120.577 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:20074-80-0
Formula:C₅H₉ClO
MDL Number:MFCD09753736