Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%
tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%
Catalog # APOSOR924855-25G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%
Catalog # APOSOR924855-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :213.23 g/mol
Storage Temperature:Ambient
CAS Number:396731-40-1
Formula:C₁₀H₁₅NO₄
MDL Number:MFCD14706038