1-(2-Chlorophenyl)-2-methylpropan-2-ol 95%
Catalog # APOSOR923685-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.663 g/mol
Storage Temperature:Ambient
CAS Number:6256-31-1
Formula:C₁₀H₁₃ClO
MDL Number:MFCD11610689