Methyl[(1-methyl-1H-pyrazol-4-yl)methyl]amine dihydrochloride 95%
Catalog # APOSOR921502-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.094 g/mol
Storage Temperature:Ambient
CAS Number:1431965-92-2
Formula:C₆H₁₁N₃·2ClH
MDL Number:MFCD25371024