Order Entry
Germany
ContactUsLinkComponent
5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester 95%
5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester 95%
Catalog # APOSOR921219-1G
Supplier:  Apollo Scientific
CAS Number:  
5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester 95%
Catalog # APOSOR921219-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.204 g/mol
Storage Temperature:Ambient
CAS Number:139183-98-5
Formula:C₉H₁₂N₂O₂
MDL Number:MFCD11849321