Ethyl tetrahydro-2-furoate 95%
Catalog # APOSOR913297-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.168 g/mol
Storage Temperature:Ambient
CAS Number:16874-34-3
Formula:C₇H₁₂O₃
MDL Number:MFCD08063871