Order Entry
Germany
ContactUsLinkComponent
5-Methylindole-3-carboxylic acid methyl ester 95%
5-Methylindole-3-carboxylic acid methyl ester 95%
Catalog # APOSOR912748-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Methylindole-3-carboxylic acid methyl ester 95%
Catalog # APOSOR912748-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :189.211 g/mol
Storage Temperature:Ambient
CAS Number:227960-12-5
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD02179412