Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 3-bromoindazole-1-carboxylate
tert-Butyl 3-bromoindazole-1-carboxylate 97%
tert-Butyl 3-bromoindazole-1-carboxylate 97%
APOSOR910593-1G
:
Apollo Scientific
:
tert-Butyl 3-bromoindazole-1-carboxylate 97%
APOSOR910593-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
297.148 g/mol
:
Refrigerator
:
1257296-40-4
:
C₁₂H₁₃BrN₂O₂
:
MFCD22380231