Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
methyl tetrahydro-2H-pyran-2-carboxylate
methyl tetrahydro-2H-pyran-2-carboxylate 98%
methyl tetrahydro-2H-pyran-2-carboxylate 98%
:
Apollo Scientific
methyl tetrahydro-2H-pyran-2-carboxylate 98%
:
Apollo Scientific
:
84355-44-2
:
:
C₇H₁₂O₃
:
MFCD12198853
:
144.168 g/mol
:
Ambient