Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-butyl morpholine-4-carboxylate
tert-butyl morpholine-4-carboxylate 98%
tert-butyl morpholine-4-carboxylate 98%
tert-butyl morpholine-4-carboxylate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
tert-butyl morpholine-4-carboxylate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
220199-85-9
:
Formula:
C₉H₁₇NO₃
MDL Number:
MFCD11111782
:
187.236 g/mol
:
GHS07
Storage Temperature:
Ambient