Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-4-methylpyridin-2-ol
5-Chloro-4-methylpyridin-2-ol
5-Chloro-4-methylpyridin-2-ol
:
Apollo Scientific
5-Chloro-4-methylpyridin-2-ol
:
Apollo Scientific
:
886364-92-7
:
:
C₆H₆ClNO
:
MFCD07375093
:
143.571 g/mol
:
GHS07
:
Ambient