Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(But-2-yl)piperazine
1-(But-2-yl)piperazine 98%
1-(But-2-yl)piperazine 98%
1-(But-2-yl)piperazine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(But-2-yl)piperazine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
34581-21-0
:
Formula:
C₈H₁₈N₂
MDL Number:
MFCD02093552
:
142.242 g/mol
Boiling Pt:
80 °C (15 mm Hg)
:
GHS07
Storage Temperature:
Ambient