Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
2'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
2'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
Irritant
2'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
675596-30-2
:
Formula:
C₁₃H₉ClO
MDL Number:
MFCD04039057
:
216.663 g/mol
:
GHS07
Storage Temperature:
Ambient