Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Benzylazetidine-2-carboxylic acid
1-Benzylazetidine-2-carboxylic acid
1-Benzylazetidine-2-carboxylic acid
1-Benzylazetidine-2-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
1-Benzylazetidine-2-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
18085-40-0
:
Formula:
C₁₁H₁₃NO₂
MDL Number:
MFCD04038976
:
191.226 g/mol
:
GHS07
Storage Temperature:
Ambient