Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-amino-3,5-dibromobenzoate
Methyl 4-amino-3,5-dibromobenzoate
Methyl 4-amino-3,5-dibromobenzoate
Irritant
Methyl 4-amino-3,5-dibromobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 4-amino-3,5-dibromobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
3282-10-8
:
Formula:
C₈H₇Br₂NO₂
MDL Number:
MFCD08436085
:
308.955 g/mol
Melting Pt:
102…104 °C
:
GHS07
Storage Temperature:
Ambient