
4-[5-(Methoxycarbonyl)pyridin-2-yl]piperazine, N1-BOC protected
Catalog # APOSOR61397-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:321.372 g/mol
Melting Pt:136…138 °C
:GHS07
Storage Temperature:Ambient
CAS Number:857284-21-0
Formula:C₁₆H₂₃N₃O₄
MDL Number:MFCD19381738