3-(4-Bromophenyl)oxetan-3-amine hydrochloride
Catalog # APOSOR61249-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.547 g/mol
Melting Pt:234…236 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1349718-53-1
Formula:C₉H₁₀BrNO·ClH
MDL Number:MFCD19687804