Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trichlorobenzaldehyde
2,4,5-Trichlorobenzaldehyde
2,4,5-Trichlorobenzaldehyde
Corrosive
Irritant
2,4,5-Trichlorobenzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2,4,5-Trichlorobenzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
35696-87-8
:
Formula:
C₇H₃Cl₃O
MDL Number:
MFCD09999820
ADR:
8,III
:
209.457 g/mol
UN:
1759
:
GHS05
:
GHS07
Storage Temperature:
Ambient