Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloropyridine-2-carboxamide
6-Chloropyridine-2-carboxamide 97%
6-Chloropyridine-2-carboxamide 97%
6-Chloropyridine-2-carboxamide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloropyridine-2-carboxamide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
70593-61-2
:
7059-61-2
Formula:
C₆H₅ClN₂O
MDL Number:
MFCD00708892
:
156.569 g/mol
:
GHS07
Storage Temperature:
Ambient